missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonic Acid, 98+%, for Protein Hydrolysis with Reduced TRP Losses, Fisher Chemical

Product Code. 10151670 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic Coated Glass bottle
Unit Size:
100g
Product Code. Quantity Packaging unitSize
10151670 100 g Plastic Coated Glass bottle 100g
1 options
This item is not returnable. View return policy

Product Code. 10151670

Brand: Fisher Chemical M/3874/48

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 75-75-2
Melting Point 19°C
Color Colorless
pH 1
Boiling Point 167°C
Packaging Plastic Coated Glass bottle
Molecular Formula CH4O3S
MDL Number 7518
Quantity 100 g
Synonym methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane
InChI Key AFVFQIVMOAPDHO-UHFFFAOYSA-N
SMILES CS(=O)(=O)O
IUPAC Name methanesulfonic acid
Molecular Weight (g/mol) 96.1
PubChem CID 6395
ChEBI CHEBI:27376
Formula Weight 96.1g/mol
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methanesulfonic acid
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P361-Take off immediately all contaminated clothing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.