missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonic acid, 99%, extra pure

Product Code. 10336910 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
250mL
5 L
5mL
This item is not returnable. View return policy

Product Code. 10336910

Brand: Thermo Scientific Acros 125610050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-75-2 | CH4O3S | 96.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-75-2
Molecular Formula CH4O3S
Molecular Weight (g/mol) 96.1
MDL Number MFCD00007518
InChI Key AFVFQIVMOAPDHO-UHFFFAOYSA-N
Synonym methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane
PubChem CID 6395
ChEBI CHEBI:27376
IUPAC Name methanesulfonic acid
SMILES CS(=O)(=O)O

Specifications

Melting Point 19.0°C
Density 1.4810g/mL
Boiling Point 167.0°C (10.0 mmHg)
Flash Point 189°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3SO3H
Refractive Index 1.4252 to 1.4315
Quantity 5 mL
Beilstein 04, 4
Fieser 01,666; 02,270; 04,326; 10,256; 11,321; 12,212; 13,176
Merck Index 15, 6026
Specific Gravity 1.481
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and ethanol, insoluble in hexane and methylcyclopentane, 1.50wt% benzene -0.23wt% 0-chlorotoluene (26-28°C), 0.38wt% toluene -0.47wt% ethyl disulfide (26-28°C)
Viscosity 11.6 mPa.s (25°C)
Formula Weight 96.1
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Methanesulfonic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.