missing translation for 'onlineSavingsMsg'
Learn More

Methanol with 0.1% acetic acid, Honeywell™

Product Code. 15604340
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15604340 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15604340 Supplier Honeywell Riedel-de Haën Supplier No. 346722.5L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 67-56-1
Quantity 2.5 L
Flash Point 11°C (51.8°F)
Molecular Formula CH4O
MDL Number MFCD00004595
UN Number UN1230
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
PubChem CID 887
ChEBI CHEBI:17790
Formula Weight 32.04g/mol
Percent Purity ≥99.5% (GC)
Chemical Name or Material Methanol with 0.1% acetic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.