missing translation for 'onlineSavingsMsg'
Learn More

Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur.

Product Code. 10607221
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Metal Drum
Unit Size:
1 L
2.5 L
25 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10607221 500 mL EcoSafPak™ Glass Bottle 500mL
10107660 1 L EcoSafPak™ Glass Bottle 1 L
10073023 2.5 L Glass Bottle 2.5 L
10668323 25 L Metal Drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10607221 Supplier GC Internal (Acros/Geel) Supplier No. 423955000
  • ONLINE EXCLUSIVE PRICE
    27.55 €
    26.70 € / 500mL
    Save 0.85 €
    3% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 67-56-1 | CH4O | 32.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98.0°C
Density 0.7910g/mL
Quantity 500 mL
Flash Point 12°C
Packaging EcoSafPak™ Glass Bottle
Physical Form Liquid
Refractive Index 1.3280 to 1.3300
Residue after Evaporation 0.001% max.
Assay Percent Range 99.8+%
Specific Gravity 0.791
Titratable Acid 0.0003 meq/g max.
Titratable Base 0.0002 meq/g max.
Linear Formula CH3OH
Vapor Pressure Vapor Pressure: 128hPa at 20°C
Viscosity 0.55 mPa.s (20°C)
Water 0.1% max. (K.F.)
Beilstein 01, IV, 1227
Fieser 11,322; 13,337
Merck Index 15, 6029
Boiling Point 64.7°C
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
Formula Weight 32.04
Percent Purity ≥99.8%
Grade ACS
Chemical Name or Material Methanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P330-Rinse mouth.
  • P363-Wash contaminated clothing before reuse.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: methanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.