missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, 99.8%, for residue analysis, ECD tested for pesticide analysis
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For residue analysis
Specifications
Specifications
| Melting Point | -98.0°C |
| Density | 0.7910g/mL |
| Boiling Point | 64.7°C |
| Quantity | 1 L |
| Flash Point | 12°C |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Refractive Index | 1.3280 to 1.3300 |
| Assay Percent Range | 99.8% |
| Residue after Evaporation | 0.0002% max. |
| Show More |
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?