missing translation for 'onlineSavingsMsg'
Learn More

Methanol, ≥99.8%, for spectroscopy ACS

Product Code. 10209370 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Unit Size:
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 10209370

Brand: Thermo Scientific Acros 413775000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 67-56-1 | CH4O | 32.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98.0°C
Absorbance 0.01 max. at 280nm, 0.01 max. at 400nm, 0.04 max. at 260nm, 0.1 max. at 240nm, 0.2 max. at 230nm, 0.4 max. at 220nm, 0.8 max. at 210nm, 1 max. at 205nm
Density 0.7910g/mL
Boiling Point 64.7°C
Quantity 500 mL
Flash Point 12°C
Packaging EcoSafPak™ Glass Bottle
Physical Form Liquid
Refractive Index 1.3280 to 1.3300
Assay Percent Range 99.8%
Residue after Evaporation 0.001% max.
Specific Gravity 0.791
Titratable Acid 0.0003 meq/g max.
Linear Formula CH3OH
Titratable Base 0.0002 meq/g max.
Vapor Pressure Vapor Pressure: 128hPa at 20°C
Viscosity 0.55 mPa.s (20°C)
Beilstein 01, IV, 1227
Water 0.1% max. (K.F.)
Fieser 11,322; 13,337
Merck Index 15, 6029
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
Formula Weight 32.04
Percent Purity 99.8%
Grade ACS
Chemical Name or Material Methanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.