missing translation for 'onlineSavingsMsg'
Learn More

Methanol, 99.8%, for HPLC
GREENER_CHOICE

Product Code. 10285131 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10285131 1 L Glass Bottle 1 L
10124490 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10285131 Supplier Thermo Scientific Supplier No. 268280010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For HPLC | CAS: 67-56-1 | CH4O | 32.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98.0°C
Absorbance (1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.005 max. at 250nm, 0.005 max. at 260nm, 0.02 max. at 240nm, 0.05 max. at 230nm, 0.1 max. at 220nm, 0.2 max. at 210nm, 1 max. at 205nm
Color Colorless to Yellow
Acidity 0.0002 meq/g max.
Alkalinity 0.0002 meq/g max.
Density 0.7910g/mL
Quantity 1 L
Flash Point 12°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3280 to 1.3300
Residue after Evaporation 5ppm max.
Assay Percent Range 99.8%
Specific Gravity 0.791
Linear Formula CH3OH
Vapor Pressure Vapor Pressure: 128hPa at 20°C
Viscosity 0.55 mPa.s (20°C)
Water 0.05% Max.
Beilstein 01, IV, 1227
Fieser 11,322; 13,337
Merck Index 15, 6029
Green Features ACT Ecolabel (Silver)
Boiling Point 64.7°C
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
Formula Weight 32.04
Percent Purity 99.8%
Grade HPLC
Chemical Name or Material Methanol, for HPLC
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.