missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, 99.9%, for biochemistry, AcroSeal™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
1 L
2.5 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
Specifications
Specifications
| Melting Point | -98.0°C |
| Absorbance | 0.01 max. at 260nm, 0.01 max. at 400nm, 0.05 max. at 240nm, 0.15 max. at 230nm, 0.3 max. at 220nm, 0.6 max. at 210nm, 1 max. at 207nm |
| Density | 0.7910g/mL |
| Boiling Point | 64.7°C |
| Quantity | 2.5 L |
| Flash Point | 12°C |
| Packaging | AcroSeal™ Glass Bottle |
| Refractive Index | 1.3280 to 1.3300 |
| Assay Percent Range | 99.9% |
| Specific Gravity | 0.791 |
| Show More |
Safety and Handling
Product Identifier
- Methanol
- Danger
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- MIXTURE LIST-Contains : methanol
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction