missing translation for 'onlineSavingsMsg'
Learn More

Methanol, ≥99.9%, Solstice TraceSELECT™

Product Code. 15695480
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15695480 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15695480 Supplier Honeywell-Riedel-de Haen Supplier No. 421051L
 Restricted Item
Estimated Shipment: 27-05-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TraceSELECT™, for metal speciation analysis,≥99.9%

  • High purity solvents for speciation analysis by LC-ICP-MS

  • Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance

  • The blank values for metal traces in these solvents are in the ppb range or lower.

TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Density 0.791g/mL at 25°C
Quantity 1 L
Explosion Limit 0.36
Flash Point 9°C (48.2°F)
Ignition Point 725°F
Packaging Glass Bottle
Molecular Formula CH4O
Linear Formula CH3OH
Vapor Pressure 410mmHg (50°C), 97.68mmHg (20°C)
MDL Number MFCD00004595
UN Number UN1230
Beilstein 1098229
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
PubChem CID 887
ChEBI CHEBI:17790
Formula Weight 32.04g/mol
Percent Purity ≥99.9%
Grade TraceSELECT
Chemical Name or Material Methanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.