missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Absolute-Acetone Free, Solstice

Product Code. 15697930
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
Unit Size:
1 GAL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15697930 4 L Glass Bottle 1 GAL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15697930 Supplier Fluka Supplier No. M17751GA
  • ONLINE EXCLUSIVE PRICE
    146.00 €
    119.18 € /
    Save 26.82 €
    18% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 67-56-1
Density 0.791g/mL at 25°C
Quantity 4 L
Explosion Limit 0.36
Flash Point 9°C (48.2°F)
Ignition Point 725°F
Packaging Glass Bottle
Molecular Formula CH4O
Linear Formula CH3OH
Vapor Pressure 410mmHg (50°C), 97.68mmHg (20°C)
MDL Number MFCD00004595
UN Number UN1230
Beilstein 1098229
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
PubChem CID 887
ChEBI CHEBI:17790
Formula Weight 32.04g/mol
Chemical Name or Material Methanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.