missing translation for 'onlineSavingsMsg'
Learn More

Methanol, ACS reagent, ≥99.8%, Solstice

Product Code. 15671610
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
6 x 500 mL
20 L
6 x 1 L
Unit Size:
1 L
2.5 L
20 L
4 L
500mL
Pack of 6
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15671610 500 mL 500mL
15641610 1 L 1 L
15651610 2.5 L 2.5 L
15691610 6 x 500 mL Pack of 6
15678320 4 L 4 L
15681610 6 x 1 L Pack of 6
15661610 20 L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15671610 Supplier Honeywell-Riedel-de Haen Supplier No. 179337500ML
  • ONLINE EXCLUSIVE PRICE
    42.00 €
    30.85 € / 500mL
    Save 11.15 €
    26% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS reagent,≥99.8%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C (literature)
CAS Max % 100.0000%
Density 0.791 g/mL at 25 °C(lit.)
Quantity 500 mL
Flash Point 11°C
Packaging Glass Bottle
Linear Formula CH3OH
UN Number UN1230
Beilstein 1098229
Boiling Point 64.7°C
Formula Weight 32.04
Percent Purity >99.8%
Grade ACS
Chemical Name or Material Methanol, ACS reagent, ≥99.8%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301+H311+H331-Toxic if swallowed, in contact with skin or if inhaled
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.