missing translation for 'onlineSavingsMsg'
Learn More

Methanol, AMD CHROMASOLV™, ≥99.9%, Solstice

Product Code. 15604700
Change view
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
Unit Size:
2.5 L
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15604700 4 x 2.5 L Glass Bottle Pack of 4
15694690 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15604700 Supplier Honeywell-Chromasolv Supplier No. 348984X2.5L
  • ONLINE EXCLUSIVE PRICE
    306.00 €
    250.80 € / Pack of 4
    Save 55.20 €
    18% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHROMASOLV™ AMD,≥99.9%

  • Solvents designed for high-performance thin-layer chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C (literature)
CAS Max % ≤100.0000%
Density 0.791 g/mL at 25 °C(lit.)
Quantity 4 x 2.5 L
Flash Point 11°C
Packaging Glass Bottle
Linear Formula CH3OH
UN Number UN1230
Beilstein 1098229
Boiling Point 64°C to 65°C
Formula Weight 32.04
Percent Purity >99.9%
Grade High Purity
Chemical Name or Material Methanol, AMD CHROMASOLV™, ≥99.9%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.