missing translation for 'onlineSavingsMsg'
Learn More

Methanol, BAKER ANALYZED HPLC, J.T. Baker™

Catalog Number 10241061
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
EcoTainer Metal Safety Can
Glass
This item is not returnable. View return policy
This item is not returnable. View return policy

Get rapid, reproducible performance and separation results. Methanol, BAKER ANALYZED HPLC, J.T. Baker™ is an application-optimized, function-tested HPLC reagent/solvent that maximizes throughput and ease-of-use without sacrificing efficiency or reproducibility.

BAKER ANALYZED HPLC Reagents are high purity solvents, buffer salts, and mobile phase modifiers for use in analytical and preparative separations. They are optimized for research and quality control applications, including high performance liquid chromatography, gel permeation chromatography and UV-spectrophotometric analysis.

  • Recalibration or adjustments due to lot changes are minimized for optimum performance from sensitive instrumentation
  • Low backgrounds free of extraneous peaks
  • Low UV absorbance in critical ranges - assured through UV absorbance testing at a variety of points and through gradient elution testing
  • Fluorescence testing for trace impurities which would cause interference
  • Function-tested for: assay, water, minimal residue after evaporation and UV absorbance and fluorescence in critical ranges
  • Lot to lot consistency, with an actual Lot Analysis on the label
  • Innovative packaging options to assure solvent quality to the point-of-use
Melting Point -97.8°C
Absorbance Max. 0.30 at 225nm, Max. 0.10 at 240nm, Max. 0.01 at 265nm
Color Colorless
CAS 67-56-1
Molecular Formula CH4O
Linear Formula CH3OH
Synonym Carbinol, Methyl alcohol, Wood alcohol
Formula Weight 32.04g/mol
Percent Purity ≥99.8%
Grade HPLC Isocratic Grade
Density 0.79 g/mL at 25°C
Boiling Point 64°C
Quantity 1 L
Explosion Limit 6 - 36 Vol %
Flash Point 11°C
Ignition Point 240°C
Odor Alcohol
Packaging Glass
Physical Form Liquid
Refractive Index 1.3314 nD
Residue after Evaporation Max. 5 ppm
Titratable Acid Max. 0.0003meq/g
Titratable Base Max. 0.0002meq/g
Vapor Pressure 16.9 kPa at 25°C
Water Max. 0.05%
Chemical Name or Material Methanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'