missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, BAKER ANALYZED, J.T. Baker™
Click to view available options
Quantity:
2.5 L
Packaging:
Glass
Description
J.T. Baker™ spectrometry solvents are manufactured to absolutely minimize lot-to-lot variability and contaminants that can interfere with wavelengths being measured.
- 50% to 100% transmittance windows in IR wavelengths reported
- Low UV absorbance and optical absorbance
- Extremely low residue after evaporation, low acid and base concentrations
- Specifications that exceed ACS standards
- Reliable, consistent results
- Affordable pricing
Specifications
Melting Point | -97.8°C |
Absorbance | Max. 1.00 at 205nm, Max. 0.80 at 210nm, Max. 0.40 at 220nm, Max. 0.20 at 230nm, Max. 0.10 (at 240nm) - 0.01 (at 280-400nm) |
CAS | 67-56-1 |
Color | Colorless |
Molecular Formula | CH4O |
Linear Formula | CH3OH |
MDL Number | MFCD00004595 |
Appearance | Clear |
Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
Solubility Information | Miscible in ether, benzene, ethanol; Soluble in acetone, chloroform |
Show More |
Safety and Handling
Product Identifier
- Methanol
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'