missing translation for 'onlineSavingsMsg'
Learn More

Methanol, BAKER ANALYZED Pesticide Reagent, J.T. Baker™

Product Code. 15263498 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15263498 2.5 L Glass 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15263498 Supplier JT Baker Supplier No. 5279.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specially developed for use in evaluation of pesticide and insecticide residues in food, feed, water or soil samples. Methanol, BAKER ANALYZED Pesticide Reagent, J.T. Baker™ is suitable for all analysis of common pesticides.

J.T. Baker BAKER ANALYZED Pesticide Reagent Solvents:

  • Suitable for analysis of most common pesticides, such as 2-chlorobiphenyl , Heptachlor, Aldrin, Parathion, Dieldrin, Endrin, DDT, Dioxine, etc.
  • Specified to the required low residue levels (5 ppm) for use in research and quality control laboratories
  • Ideal for gas chromatography applications, testing for ECD and/or FID sensitive impurities, organic residues, and residue after evaporation

Specifications

Melting Point -98°C
CAS 67-56-1
Color Colorless
Acidity Max. 0.0005meq/g
Density 0.79g/cm3 at 20°C
Quantity 2.5 L
Explosion Limit 5.5 - 44 Vol %
Flash Point 11°C
Ignition Point 455°C
Odor Alcohol-like
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 5 ppm
Molecular Formula CH4O
Linear Formula CH3OH
MDL Number MFCD00004595
Water Max. 0.02%
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
Boiling Point 64.7°C
Solubility Information Fully miscible in water
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
PubChem CID 887
ChEBI CHEBI:17790
Formula Weight 32.04g/mol
Percent Purity Min. 99.9%
Grade Pesticide
Chemical Name or Material Methanol
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Methanol
Signalwort
  • Danger
Gefahrenkategorie
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Gefahrenhinweis
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.