missing translation for 'onlineSavingsMsg'
Learn More

Methanol, BAKER ANALYZED™, UHPLC, J.T. Baker™

Product Code. 10007540 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10007540 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10007540 Supplier JT Baker Supplier No. 8402.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gradient grade for HPLC

  • Fluorescence Trace Impurities (as quinine base), ppb:
    • Measured at Emission Maximum for Solvent Impurities max. 1.0
    • Measured at 450 nm max. 0.3
    • Gradient Elution Test Ultraviolet Absorbance (a.u.):
      • at 235 nm max. 0.005
      • at 254 nm max. 0.001
    • Ultraviolet Absorbance (1.00-cm path vs water):
      • at 225 nm max. 0.20
      • at 254 nm max. 0.02
      • at 280 nm max. 0.01
      • at 350 nm max. 0.01
      • UV Cut-off, nm max. 206

Specifications

Melting Point -98°C
CAS 67-56-1
Density 0.7918 g/cm³ (20°C)
Quantity 1 L
Flash Point 11°C
Packaging Plastic Bottle
Residue after Evaporation max. 2 ppm
Assay Percent Range min. 99.8%
Titratable Acid max. 0.0003 meq/g
Molecular Formula CH4O
Titratable Base max. 0.0001 meq/g
MDL Number MFCD00004595
Water max. 0.02%
Boiling Point 64.6°C (1013 hPa)
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.