missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Capillary GC Grade; ≥99.9%, Honeywell Riedel-de Haën™

Product Code. 15625460
Change view
Click to view available options
Quantity:
4 L
4 x 4 L
Packaging:
Glass Bottle
Unit Size:
4 L
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15625460 4 x 4 L Glass Bottle Pack of 4
15658400 4 L Glass Bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15625460 Supplier Honeywell-Chromasolv Supplier No. 4147194X4L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Capillary GC grade,≥99.9%

  • For GC headspace applications
  • Meets USP, Ph. Eur. and ICH requirements
  • GC headspace application tested
  • Packed under inert gas for longer shelf life
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -95°C
Color Colorless
Density 0.791g/mL at 25°C, 0.791g/mL at 25°C
Quantity 4 x 4 L
Explosion Limit 0.36
Flash Point 9°C (48.2°F)
Ignition Point 725°F
Odor Characteristic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.329
Linear Formula CH3OH
Vapor Pressure 97.68mmHg (20°C), 410mmHg (50°C)
UN Number UN1230
Beilstein 1098229
Boiling Point 64.7°C
Formula Weight 32.04g/mol
Percent Purity ≥99.9%
Grade CHROMASOLV™ Capillary GC
Chemical Name or Material Methanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.