missing translation for 'onlineSavingsMsg'
Learn More

Methanol CHROMASOLV™, for HPLC, ≥99.9%, Solstice

Product Code. 15604580
Change view
Click to view available options
Quantity:
1 L
2 L
2.5 L
2.5 L-PC
4 x 2 L
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2 L
2.5 L
Pack of 4
Pack of 6
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15604580 6 x 1 L Pack of 6
15644570 1 L 1 L
15674570 2 L 2 L
15654570 2.5 L 2.5 L
15664570 2.5 L-PC 2.5 L
15694570 4 x 2 L Pack of 4
15684570 4 x 2.5 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15604580 Supplier Honeywell-Chromasolv Supplier No. 348606X1L
Solstice Chromasolv Promotion
Use promo code 27944
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Product filtered through a 0.2μm filter

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Density 0.791g/mL at 25°C
Quantity 6 x 1 L
Refractive Index n20/D 1.329
Linear Formula CH3OH
Vapor Pressure 97.68mmHg (20°C)
Beilstein 1098229
Boiling Point 64.7°C
Formula Weight 32.04g/mol
Percent Purity ≥99.9%
Grade CHROMASOLV
Chemical Name or Material Methanol, for HPLC
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301+H311+H331-Toxic if swallowed, in contact with skin or if inhaled
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.