missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol CHROMASOLV™, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2 L
2.5 L
2.5 L-PC
4 x 2 L
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2 L
2.5 L
Pack of 4
Pack of 6
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
| Melting Point | -98°C |
| Density | 0.791g/mL at 25°C |
| Boiling Point | 64.7°C |
| Quantity | 2.5 L-PC |
| Refractive Index | n20/D 1.329 |
| Linear Formula | CH3OH |
| Vapor Pressure | 97.68mmHg (20°C) |
| Beilstein | 1098229 |
| Formula Weight | 32.04g/mol |
| Percent Purity | ≥99.9% |
| Show More |
Safety and Handling
Product Identifier
- Methanol
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction