missing translation for 'onlineSavingsMsg'
Learn More

Methanol-d{4}, 99.8% (Isotopic), contains 0.05% v/v TMS

Product Code. 11344378
Change view
Click to view available options
Quantity:
10 Ea.
2 Ea.
Unit Size:
Pack of 10
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11344378 2 Ea. Pack of 2
11334378 10 Ea. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11344378 Supplier Thermo Scientific Alfa Aesar Supplier No. 042319.SA
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 811-98-3 | CH4O | 36.066 g/mol

Methyl alcohol-d4 is the labeled analog of methyl alcohol, a common laboratory solvent. It is a common solvent used in NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 811-98-3
Molecular Formula CH4O
Molecular Weight (g/mol) 36.066
MDL Number MFCD00044637
InChI Key OKKJLVBELUTLKV-MZCSYVLQSA-N
Synonym methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d
PubChem CID 71568
IUPAC Name trideuterio(deuteriooxy)methane
SMILES CO

Specifications

Physical Form Liquid
Color Colorless
Density 0.89 g/mL
Odor Mild
Packaging Prescored Ampule
Quantity 2 Ea.
Melting Point -98°C
Boiling Point 64.7°C
Linear Formula CD3OD
UN Number UN1230
Solubility Information Miscible with water,ethanol,ether,acetone and chloroform.
Formula Weight 36.07
Percent Purity 99.8%
Concentration or Composition (by Analyte or Components) Contains 0.05% v/v TMS
Chemical Name or Material Methanol-d4, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.