missing translation for 'onlineSavingsMsg'
Learn More

Methanol-d3, for NMR, 99.5 atom % D

Product Code. 10358830 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 mL
10 mL
Packaging:
Glass Bottle
Unit Size:
10mL
2.5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10358830 2.5 mL Glass Bottle 2.5mL
10423225 10 mL Glass Bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10358830 Supplier Thermo Scientific Acros Supplier No. 300730025
  • ONLINE EXCLUSIVE PRICE
    94.90 €
    92.10 € / 2.5mL
    Save 2.80 €
    2% Off
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 1849-29-2 | CH4O | 35.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1849-29-2
Molecular Formula CH4O
Molecular Weight (g/mol) 35.06
MDL Number MFCD00055877
InChI Key OKKJLVBELUTLKV-FIBGUPNXSA-N
Synonym methanol-d3, methan-d3-ol, 2h3 methanol, methyl-d3 alcohol, methan-d3-ol 6ci,7ci,8ci,9ci, cd3oh, 1,1,1-trideuteromethanol, methanol d, 1,1,1-2h3 methanol, methanol-d3, 99.8 atom % d
PubChem CID 123132
IUPAC Name trideuteriomethanol
SMILES CO

Specifications

Physical Form Liquid
Color Colorless
Density 0.8670g/mL
Deuterium (D) 99.4% min.
Packaging Glass Bottle
Water 0.1% max. (Coulometric)
Quantity 2.5 mL
Linear Formula CD3OH
Formula Weight 35.07
Grade NMR
Chemical Name or Material Methanol-d3, 99.5 atom % D, For NMR
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol-d3
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: (2H3)Methanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.