missing translation for 'onlineSavingsMsg'
Learn More

Methanol-d4, for NMR, 100.0 atom % D

Product Code. 16398186 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16398186 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16398186 Supplier Thermo Scientific Acros Supplier No. 186450100
  • ONLINE EXCLUSIVE PRICE
    433.00 €
    398.00 € / 10mL
    Save 35.00 €
    8% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 811-98-3 | CD4O | 36.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Density 0.8880 g/mL
Flash Point 12°C
Refractive Index 1.3262
Specific Gravity 0.888
Quantity 10 mL
CAS 811-98-3
Melting Point -99.0°C
Boiling Point 65.0°C
Molecular Formula CD4O
Linear Formula CD3OD
Beilstein 01, III, 1187
Solubility Information Solubility aq. soln.: miscible. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
Molecular Weight (g/mol) 36.06
Formula Weight 36.06
Grade NMR
Chemical Name or Material Methanol-d4
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.