missing translation for 'onlineSavingsMsg'
Learn More

Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8 atom % D

Product Code. 16328196 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
25 mL
50 mL
Packaging:
Glass Bottle
Unit Size:
10mL
25mL
50mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
16328196 25 mL Glass Bottle 25mL
10041641 5 mL Glass Bottle 5mL
10234701 10 mL Glass Bottle 10mL
10051641 50 mL Glass Bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16328196 Supplier Thermo Scientific Acros Supplier No. 329920250
  • ONLINE EXCLUSIVE PRICE
    364.00 €
    334.00 € / 25mL
    Save 30.00 €
    8% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Methanol-d4, CH4O, CAS Number-811-98-3, methan-d3-ol-d, 2h4 methanol, methyl-d3 alcohol-d, perdeuteromethanol, methanol-d4, tetradeuteromethanol, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d, methyl alcohol-d4, cd3od, 50mL, 01, III, 1187, Colorless, 0.8880g/mL, 99.75% min., 212-378-6 | CAS: 811-98-3 | CH4O | 36.066 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 811-98-3
Molecular Formula CH4O
MDL Number MFCD00044637
InChI Key OKKJLVBELUTLKV-MZCSYVLQSA-N
Synonym trideuterio deuteriooxy methane, perdeuteromethanol, methyl-d3 alcohol-d, methan-d3-ol-d, methyl alcohol-d4, tetradeuteromethanol, 2h4 methanol, cd3od, methanol-d4, methanol-d4, 99.8 atom % d
PubChem CID 71568

Specifications

Physical Form Liquid
Color Colorless
Density 0.8880 g/mL
Deuterium (D) 99.75% min.
Flash Point 12°C
Packaging Glass Bottle
Refractive Index 1.3262
Water 0.05% max. (Coulometric)
Quantity 25 mL
Melting Point -99.0°C
Boiling Point 65.0°C
Linear Formula CD3OD
Beilstein 01, III, 1187
SMILES [2H]OC([2H])([2H])[2H]
IUPAC Name (²H₃)methan(²H)ol
Molecular Weight (g/mol) 36.07
Formula Weight 36.06
Grade NMR
Chemical Name or Material Methanol-d4, 99.8 atom % D
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol-d4
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: methanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.