missing translation for 'onlineSavingsMsg'
Learn More

Methanol-d4, for NMR, packaged in 0.75 ml ampoules, 100.0 atom % D

Product Code. 10021691 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
7.5 mL
Unit Size:
7.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10021691 7.5 mL 7.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10021691 Supplier Thermo Scientific Acros Supplier No. 351460075
  • ONLINE EXCLUSIVE PRICE
    188.00 €
    182.00 € / 7.5mL
    Save 6.00 €
    3% Off
View all promotions
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 811-98-3 | CH4O | 36.066 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Color Colorless
Density 0.8880g/mL
Deuterium (D) 99.95% min.
Flash Point 12°C
Refractive Index 1.3262
Water 0.02% max. (Coulometric)
Quantity 7.5 mL
CAS 811-98-3
Melting Point -99.0°C
Boiling Point 65.0°C
Molecular Formula CH4O
Linear Formula CD3OD
MDL Number MFCD00044637
Beilstein 01, III, 1187
Synonym methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d
InChI Key OKKJLVBELUTLKV-MZCSYVLQSA-N
SMILES CO
IUPAC Name trideuterio(deuteriooxy)methane
Molecular Weight (g/mol) 36.066
PubChem CID 71568
Formula Weight 36.06
Grade NMR
Chemical Name or Material Methanol-d4, 100.0 atom% D
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol-d4
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.