missing translation for 'onlineSavingsMsg'
Learn More

Methanol, HPLC for Gradient Analysis, Fisher Chemical™

Brand:  Fisher Chemical M/4058/15

18.96 EUR valid until 2021-06-30
Use promo code "14804" to get your promotional price.



Code : 25

Additional Details : CAS Number : 67-56-1 Weight : 0.79200kg Transport : UN number : 1230 Chem class : 3 Pack group : II


Packaging Amber glass bottle
Quantity 1L

 View more versions of this product

Product Code. 10010280

Quantity Price
1 30.69 € / 1 L
4 27.62 € / 1 L
16 26.09 € / 1 L
In stock, ready to ship: 125

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specification
Molecular Formula: CH4O
Water Soluble: Yes
Molecular Weight: 32.04
Melting Point: -98°C
Health Hazards: Causes moderate eye irritation, may cause skin irritation, may cause irritation of the digestive tract
For Use With [Application]: HPLC for gradient analysis
Flash Point: 12°C
CAS No.: 67-56-1
Boiling Point: 64.7°C
Hazard Symbol: F, T
Synonyms: Carbinol, Methyl Alcohol, Methyl Hydroxide, Monohydroxymethane, Wood Alcohol, Wood Naptha, Wood Spirits, Columbian Spirits
Specifications

Chemical Identifiers

67-56-1
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
4595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO

Specifications

67-56-1
4595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04g/mol
Amber glass bottle
1.11
64.7°C
-98°C
CH4O
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CO
32.042
CHEBI:17790
Liquid
128 hPa at 20°C
0.55cp at 20°C
Colorless
1L
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methanol

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P240-Ground/bond container and receiving equipment.

SDS Documents