missing translation for 'onlineSavingsMsg'
Learn More

Methanol, HPLC, Reagent European Pharmacopoeia,Solstice

Product Code. 16685883
Change view
Click to view available options
200 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16685883 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16685883 Supplier Burdick & Jackson BioSyn Supplier No. 65725200LRC
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

Melting Point -98°C
CAS 67-56-1
Color Colorless
CAS Max % 1
Density 0.79 g/cm3 (20°C)
Quantity 200 L
Flash Point 11°C
Molecular Formula CH4O
Linear Formula CH3OH
Beilstein 1098229
Synonym Methyl alcohol
Boiling Point 64°C to 65°C (1013 hPa)
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Grade GC
Chemical Name or Material Methanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.