missing translation for 'onlineSavingsMsg'
Learn More

Methanol, LC/MS grade, Thermo Scientific
GREENER_CHOICE

Product Code. 30280930 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Unit Size:
1 L
2.50 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30280930 500 mL 500mL
30280931 1 L 1 L
30280932 2.5 L 2.50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30280930 Supplier Thermo Scientific Supplier No. T004560500
  • ONLINE EXCLUSIVE PRICE
    38.50 €
    36.30 € / 500mL
    Save 2.20 €
    5% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

Molecular Weight (g/mol) 32.04 g/mol

Specifications

Quantity 500 mL
CAS 67-56-1
Packaging Glass Bottle
Boiling Point 65°C
Percent Purity 99.90%
Grade LC-MS
Chemical Name or Material Methanol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol, LC/MS grade
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: Methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.