missing translation for 'onlineSavingsMsg'
Learn More

Methanol, LC-MS Ultra CHROMASOLV™, Solstice

Product Code. 15631400
Change view
Click to view available options
Quantity:
1 L
2 L
Unit Size:
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15631400 2 L 2 L
15621400 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15631400 Supplier Honeywell-Chromasolv Supplier No. 142622L
  • ONLINE EXCLUSIVE PRICE
    275.00 €
    136.24 € / 2 L
    Save 138.76 €
    50% Off
 Restricted Item
Estimated Shipment: 03-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHROMASOLV™ LC-MS Ultra, tested for UHPLC-MS

  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise
  • Exemplary lot-to-lot reproducibility
  • Low levels of trace metals
  • Packaged in clear borosilicate glass bottles
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Density 0.791g/mL at 25°C
Quantity 2 L
Explosion Limit 0.36
Flash Point 9°C (48.2°F)
Ignition Point 725°F
Linear Formula CH3OH
Vapor Pressure 97.68mmHg (20°C), 410mmHg (50°C)
UN Number 1230
Beilstein 1098229
Formula Weight 32.04g/mol
Grade LC-MS Ultra CHROMASOLV
Chemical Name or Material Methanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.