missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Optima™ LC/MS Grade, Fisher Chemical

Product Code. 10402824 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Unit Size:
1 L
2.5 L
4 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10402824 4 L 4 L
10636545 500 mL 500mL
10031094 1 L 1 L
10767665 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10402824 Supplier Fisher Chemical Supplier No. A4564
  • ONLINE EXCLUSIVE PRICE
    213.00 €
    206.10 € / 4 L
    Save 6.90 €
    3% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Methanol, >-99.9%, CH4O, CAS Number-67-56-1, wood alcohol, methyl hydroxide, colonial spirit, columbian spirit, wood naphtha, carbinol, methyl alcohol, pyroxylic spirit, wood spirit, methylol | CAS: 67-56-1 | CH4O | 32.04 g/mol

Methanol, >-99.9%, CH4O, CAS Number-67-56-1, wood alcohol, methyl hydroxide, colonial spirit, columbian spirit, wood naphtha, carbinol, methyl alcohol, pyroxylic spirit, wood spirit, methylol

Note: Glass bottles have fluorinated lining.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Absorbance ≤0.005 AU at 260nm, ≤0.005 AU at 280nm, ≤0.01 AU at 254nm, ≤0.1 AU at 230nm, ≤0.2 AU at 220nm, ≤0.4 AU at 214nm, ≤0.5 AU at 210nm
Color Colorless
Density 0.791g/cm³
Quantity 4 L
Flash Point 12°C (53.6°F)
Identification Pass Test
Packaging Glass Bottle with fluorinated lining
Physical Form Liquid
Residue after Evaporation ≤1ppm
Assay Percent Range Assay: 99.9% min by GC
Titratable Acid ≤0.0003meq/g
Titratable Base ≤0.0002meq/g
Linear Formula CH3OH
Vapor Pressure 128 hPa at 20°C
Viscosity 0.55cp at 20°C
UN Number 1230
Water ≤0.02%
Merck Index 15, 6029
Boiling Point 64.7°C
Formula Weight 32.04
Percent Purity ≥99.9%
Grade LC-MS & UHPLC-UV
Assay GC
Chemical Name or Material Methanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P240-Ground/bond container and receiving equipment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.