missing translation for 'onlineSavingsMsg'
Learn More

Methanol (Peroxide-Free/Sequencing), Fisher BioReagents™

Product Code. 10785484
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Unit Size:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10785484 4 L Glass Bottle 4 L
10163383 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10785484 Supplier Fisher BioReagents Supplier No. BP11054
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methanol (Peroxide-Free/Sequencing)

Methanol is used in staining and destaining protein electrophoresis gels, in HPLC, and in other biological applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Absorbance 0.025 max. at 254nm, 0.15 max. at 230nm, 0.3 max. at 220nm, 1 max. at 205nm
Color Colorless
Density 0.791g/cm³
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.326 to 1.330 (at 25°C)
Residue after Evaporation 3ppm max.
Assay Percent Range ≥99.9 %
Titratable Acid 0.0003mEq/g max.
Titratable Base 0.0002mEq/g max.
Linear Formula CH3OH
Vapor Pressure 128 hPa at 20°C
Viscosity 0.55 cP at 20°C
Water 0.1% max.
Merck Index 15, 6029
Boiling Point 64.7°C
Formula Weight 32.04
Percent Purity ≥99.9%
Grade Peroxides: ≤0001%
Chemical Name or Material Methanol, Peroxide-free
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P240-Ground/bond container and receiving equipment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.