missing translation for 'onlineSavingsMsg'
Learn More

Methanol, puriss., Solstice
BIOPHARMA

Brand:  Fluka 24229-160KG

1553.08 EUR valid until 2026-12-31
BEST PRICE on Promo! Use promo code "28170" to get your promotional price.



 View more versions of this product

Product Code. 15632580

  • ONLINE EXCLUSIVE PRICE
    1635.00 €
    1343.67 € / 160kg
    Save 291.33 €
    17% Off
View all promotions
 Restricted Item
Estimated Shipment: 09-11-2026
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methanol

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301+H311+H331-Toxic if swallowed, in contact with skin or if inhaled
  • H370-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS Documents

Documents

Certificates
Promotions

Promotions

Promo
Expires: 2026-12-31
Discount Lab Supplies
Promo Code: 28170
Shop discounted lab supplies across top categories and brands.

Applying Promo and Adding

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.