missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
2.5L, Amber glass bottle
Specifications
Specifications
| Melting Point | -98°C |
| CAS | 67-56-1 |
| Color | Colorless |
| Boiling Point | 64.7°C |
| Quantity | 2.5 L |
| Packaging | Amber Glass Bottle |
| Physical Form | Liquid |
| Molecular Formula | CH4O |
| MDL Number | MFCD00004595 |
| Vapor Pressure | 128 hPa at 20°C |
| Show More |
Safety and Handling
Product Identifier
- Methanol
- Danger
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P240-Ground/bond container and receiving equipment.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?