missing translation for 'onlineSavingsMsg'
Learn More

Methapyrilene, TRC

Product Code. 30137658
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137658 1 g 1g
30009666 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137658 Supplier Toronto Research Chemicals Supplier No. TRCM2599981G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-80-5
Molecular Formula C14 H19 N3 S
Molecular Weight (g/mol) 261.39
Synonym 1,2-Ethanediamine, N1,N1-dimethyl-N2-2-pyridinyl-N2-(2-thienylmethyl)-, 1,2-Ethanediamine, N,N-dimethyl-N'-2-pyridinyl-N'-(2-thienylmethyl)- (9CI), Pyridine, 2-[[2-(dimethylamino)ethyl]-2-thenylamino]- (8CI), 2-[[2-(Dimethylamino)ethyl]-2-thenylamino]pyridine, A 3322, AH 42, Dimethyl[2-[(pyridin-2-yl)[(thien-2-yl)methyl]amino]ethyl]amine, Histadryl, Lulamin, Metapyrilene, Methapyrilene, N,N-Dimethyl-N'-pyrid-2-yl-N'-2-thenylethylenediamine, N-(α-Pyridyl)-N-(α-thenyl)-N',N'-dimethylethylenediamine, Paradormalene, Pyrinistab, Pyrinistol, Rest-on, Restryl, Semikon, Sleepwell, Tenalin, Thenylene, Thenylpyramine, Thionylan
IUPAC Name N,N-dimethyl-N'-pyridin-2-yl-N'-(thiophen-2-ylmethyl)ethane-1,2-diamine
SMILES CN(C)CCN(Cc1cccs1)c2ccccn2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C14H19N3S/c1-16(2)9-10-17(12-13-6-5-11-18-13)14-7-3-4-8-15-14/h3-8,11H,9-10,12H2,1-2H3
Formula Weight 261.13
Percent Purity >95%
Chemical Name or Material Methapyrilene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.