missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Methionine/Choline-deficient Diet, Pellets

Product Code. p-7139898 Shop All MP Biomedicals Products
Click to view available options
Packaging Quantity:
20 kg
50 kg
Unit Size:
20kg
50kg
This item is not returnable. View return policy

Product Code. 11445365

Brand: MP Biomedicals™ 0296043920

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methionine/Choline-deficient Pellet Diet, is used to study induction of non-alcoholic steatohepatitis in experimental models and its routes of development.

  • Non-alcoholic steatohepatitis(NASH) was induced by feeding a methionine/choline-deficient diet (MCD-diet) for up to 5 weeks to mice.
  • NASH is a disease with a high incidence, difficult diagnosis, and as yet no effective treatment.
  • So, MCD-diet is used to study induction of non-alcoholic steatohepatitis in experimental models and its routes of development.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Choline Deficient Diet
Format Pellets
Packaging Quantity 20 kg
Storage Requirements Store at Room Temperature (15°C to 30°C)

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.