missing translation for 'onlineSavingsMsg'
Learn More

Methotrexate Disodium Salt, TRC

Product Code. 30477745
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477745 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477745 Supplier Toronto Research Chemicals Supplier No. TRCM260753100MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methotrexate Disodium Salt
CAS 7413-34-5
Molecular Formula C20H20N8Na2O5
Quantity 100 mg
Synonym Amethopterin Sodium, Methotrexate Lederle, Methotrexate Disodium Salt, Metoart, Metotrexato Lederle, Sodium Methotrexate, N-[4-[[(2,4-Diamino-6-pteridinyl)methyl]methylamino]benzoyl]-L-glutamic Acid Disodium Salt
InChI Formula InChI=1S/C20H22N8O5.2Na/c1-28(9-11-8-23-17-15(24-11)16(21)26-20(22)27-17)12-4-2-10(3-5-12)18(31)25-13(19(32)33)6-7-14(29)30;;/h2-5,8,13H,6-7,9H2,1H3,(H,25,31)(H,29,30)(H,32,33)(H4,21,22,23,26,27);;/q;2*+1/p-2/t13-;;/m0../s1
SMILES NC1=NC(N)=NC2=C1N=C(CN(C)C3=CC=C(C(NC(C(O[Na])=O)CCC(O[Na])=O)=O)C=C3)C=N2
IUPAC Name disodium;(2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioate
Molecular Weight (g/mol) 498.4
Formula Weight 498.1352
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.