missing translation for 'onlineSavingsMsg'
Learn More

Methotrexate, MP Biomedicals™

Product Code. 11453530 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453530 500 mg 500mg
11423530 10 mg 10mg
11443530 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453530 Supplier MP Biomedicals Supplier No. 0210229990

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Acts as a folic acid antagonist. Methotrexate, MP Biomedicals is a mix of 4-amino-10-methylfolic acid and related compounds, structurally similar to folic acid. A potent anticancer agent, it blocks DNA synthesis by blocking production of tetrahydrofolate

Methotrexate is actively transported into cells by the folate transporter. In the cell, it is converted to a high molecular weight polyglutamate metabolite by folypolyglutamate synthase that, in turn, binds to dihydrofolate reductase and inhibits its activity. Methotrexate-polyglutamate is degraded intracellularly by gamma-glutamyl hydrolase.

Methotrexate is an allosteric inhibititor of dihydrofolate reductase (DHFR), the enzyme that catalyzes the conversion of dihydrofolate to tetrahydrofolate. Since tetrahydrolfolate is required for purine and pyrimidine synthesis, methotrexate treatment results in the inhibition of DNA and RNA synthesis.

  • Potent inhibitor of dihydrofolate reductase in DHFR-based protein expression systems
  • Anti-cancer agent used in antitumor studies
  • Effective in treatment of pyrimethamine-resistant Plasmodium vivax malaria parasites
  • Shows immunosuppressive effects, e.g. in rheumatoid arthritis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-05-2
Molecular Formula C20H22N8O5
Molecular Weight (g/mol) 454.447
InChI Key FBOZXECLQNJBKD-ZDUSSCGKSA-N
Synonym methotrexate, abitrexate, rheumatrex, amethopterin, mexate, metatrexan, hdmtx, trexall, methylaminopterinum, antifolan
PubChem CID 126941
ChEBI CHEBI:44185
IUPAC Name (2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid
SMILES CN(CC1=CN=C2C(=N1)C(=NC(=N2)N)N)C3=CC=C(C=C3)C(=O)NC(CCC(=O)O)C(=O)O

Specifications

Melting Point 185°C to 204°C
Boiling Point 184°C to 204°C (monohydrate decomposition)
Quantity 500 mg
Solubility Information Soluble in alkaline solutions with decomposition.Insoluble in water, ethanol, chloroform and ether.
Optical Rotation 19–24°
Formula Weight 454.4
Grade USP
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.