missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Methotrexate

Product Code. 18712724
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18712724 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18712724 Supplier Tocris Bioscience™ Supplier No. 1230/100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cytotoxic agent

Cytotoxic agent. Inhibits thymidylate synthetase and de novo purine synthesis. Potent folic acid antagonist; inhibits dihydrofolate reductase. Also inhibits Ras carboxyl methylation in DKOB8 cells, leading to decreased p44 and Akt activation.
TRUSTED_SUSTAINABILITY

Specifications

CAS 59-05-2
Quantity 100 mg
Molecular Formula C20H22N8O5
Formula Weight 454.44
Purity >99%
Molecular Weight (g/mol) 454.447
Chemical Name or Material Methotrexate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.