missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Methoxy-X04

Description
Methoxy-X04 is a fluorescent amyloid β (Aβ) probe for the detection and quantification of plaques, tangles and cerebrovascular amyloid. Methoxy-X04 Displays high in vitro binding affinity (Ki = 26.8 nM); binds selectively to fibrillar β-sheet deposits. Methoxy-X04 is a derivative of Congo Red (Cat. No. 5167). Methoxy-X04 can be used for in vivo Aβ plaque labeling. Methoxy-X04 co-localizes with CD68+ phagosomes in plaque-associated Iba1+ microglia; and it labels molecularly distinct plaque-associated microglia populations. Brain penetrant.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 344.4 |
| Purity | >98% |
| Molecular Weight (g/mol) | 344.41 |
| Chemical Name or Material | Methoxy-X04 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?