missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methoxyacetic acid, 97+%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
25mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 7°C |
| Color | Colorless to Yellow |
| Density | 1.1740g/mL |
| Boiling Point | 202°C to 204°C |
| Flash Point | 121°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97+% |
| Packaging | Glass bottle |
| Linear Formula | CH3OCH2CO2H |
| Refractive Index | 1.4153 to 1.4173 |
| Show More |
Safety and Handling
Product Identifier
- Methoxyacetic acid
- Danger
- Acute toxicity Category 4
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H360FD-May damage fertility. May damage the unborn child.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction