missing translation for 'onlineSavingsMsg'
Learn More

Methoxyamine Hydrochloride, MP Biomedicals™

Product Code. 11487692 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487692 25 g 25g
11467692 1 g 1g
11477692 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487692 Supplier MP Biomedicals Supplier No. 0215540525
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Methoxyamine Hydrochloride is used as a reagent for the preparation of O-methyl oximes.

  • Also used to make O-methyl oximes from aldehydes, or ketones.

Chemical Identifiers

CAS 593-56-6
Molecular Formula CH6ClNO
Molecular Weight (g/mol) 83.515
InChI Key XNXVOSBNFZWHBV-UHFFFAOYSA-N
Synonym o-methylhydroxylamine hydrochloride, methoxyamine hydrochloride, methoxylamine hydrochloride, methoxyammonium chloride, o-methylhydroxylamine hcl, aminooxy methane hydrochloride, methoxyamine hcl, unii-203546olmf, hydroxylamine, o-methyl-, hydrochloride, o-methylhydroxyamine hydrochloride
PubChem CID 521874
IUPAC Name O-methylhydroxylamine;hydrochloride
SMILES CON.Cl

Specifications

Melting Point 300°F to 304°F (decomposes)
Quantity 25 g
Solubility Information Soluble
Formula Weight 83.52
Physical Form Crystalline Powder
Chemical Name or Material Methoxyamine Hydrochloride

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.