missing translation for 'onlineSavingsMsg'
Learn More

Methoxycarbonylsulfenyl chloride, 97%

Product Code. 10135470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
Packaging:
Glass bottle
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10135470 1 mL Glass bottle 1mL
10269313 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10135470 Supplier Thermo Scientific Acros Supplier No. 308840010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 26555-40-8 | C2H3ClO2S | 126.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26555-40-8
Molecular Formula C2H3ClO2S
Molecular Weight (g/mol) 126.56
MDL Number MFCD00013648
InChI Key TXJXPZVVSLAQOQ-UHFFFAOYSA-N
Synonym methoxycarbonylsulfenyl chloride, methoxycarbonyl sulfenyl chloride, s-chloro o-methyl thiocarbonate, thiocarbonic acid, anhydrosulphide with methyl thiohypochlorite, carbonothioic acid, anhydrosulfide with thiohypochlorous acid, methyl ester, acmc-209gs8, methoxycarbonylsulfenylchloride, chlorothio methoxy oxomethane, methoxy carbonylsulfenyl chloride, methoxy-carbonylsulfenyl chloride
PubChem CID 123417
IUPAC Name methyl chlorosulfanylformate
SMILES COC(=O)SCl

Specifications

Color Yellow
Density 1.3900g/mL
Boiling Point 133°C to 134°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3O2CSCl
Refractive Index 1.4810 to 1.483
Quantity 1 mL
Specific Gravity 1.39
Formula Weight 126.56
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methoxycarbonylsulfenyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.