missing translation for 'onlineSavingsMsg'
Learn More

Methoxylamine hydrochloride, 25-30% aq. soln., Thermo Scientific™

Product Code. 11398145
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398145 100 g 100g
11388145 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398145 Supplier Thermo Scientific Alfa Aesar Supplier No. L08415.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Methoxyamine hydrochloride was used as a reagent in the preparation of O-methyl oximes and also used in the synthesis of O-methyl oximes from aldehydes2 or ketones. It is also applied as an internal standard for Prostaglandin assays by gas chromatography-mass spectrometry.

Solubility
Fully miscible in water. Soluble in DMSO and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-56-6
Molecular Formula CH6ClNO
Molecular Weight (g/mol) 83.515
MDL Number MFCD00012951
InChI Key XNXVOSBNFZWHBV-UHFFFAOYSA-N
Synonym o-methylhydroxylamine hydrochloride, methoxyamine hydrochloride, methoxylamine hydrochloride, methoxyammonium chloride, o-methylhydroxylamine hcl, aminooxy methane hydrochloride, methoxyamine hcl, unii-203546olmf, hydroxylamine, o-methyl-, hydrochloride, o-methylhydroxyamine hydrochloride
PubChem CID 521874
IUPAC Name O-methylhydroxylamine;hydrochloride
SMILES CON.Cl

Specifications

Density 1.1
Quantity 100 g
UN Number UN3265
Beilstein 3589723
Merck Index 14,5989
Solubility Information Fully miscible in water. Soluble in DMSO and methanol.
Formula Weight 83.52
Concentration or Composition (by Analyte or Components) 25 to 30% aq. Soln.
Chemical Name or Material Methoxylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.