missing translation for 'onlineSavingsMsg'
Learn More

Methoxylamine hydrochloride, 98+%

Product Code. 11459466
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459466 5 g 5g
11469466 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459466 Supplier Thermo Scientific Alfa Aesar Supplier No. A19188.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 593-56-6 | CH6ClNO | 83.515 g/mol

Methoxylamine hydrochloride is used as a reagent in the preparation of O-methyl oximes from aldehydes. It is orally bioavailable small molecule inhibitor with potential adjuvant activity. It is also used as internal standard for prostaglandin assays by gas chromatography-mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methoxylamine hydrochloride is used as a reagent in the preparation of O-methyl oximes from aldehydes. It is orally bioavailable small molecule inhibitor with potential adjuvant activity. It is also used as internal standard for prostaglandin assays by gas chromatography-mass spectrometry.

Solubility
Soluble in water, dimethyl sulfoxide and methanol.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-56-6
Molecular Formula CH6ClNO
Molecular Weight (g/mol) 83.515
MDL Number MFCD00012951
InChI Key XNXVOSBNFZWHBV-UHFFFAOYSA-N
Synonym o-methylhydroxylamine hydrochloride, methoxyamine hydrochloride, methoxylamine hydrochloride, methoxyammonium chloride, o-methylhydroxylamine hcl, aminooxy methane hydrochloride, methoxyamine hcl, unii-203546olmf, hydroxylamine, o-methyl-, hydrochloride, o-methylhydroxyamine hydrochloride
PubChem CID 521874
IUPAC Name O-methylhydroxylamine;hydrochloride
SMILES CON.Cl

Specifications

Melting Point ∼148°C (decomposition)
Quantity 5 g
UN Number UN3261
Beilstein 3589723
Sensitivity Hygroscopic
Merck Index 14,5989
Solubility Information Soluble in water,dimethyl sulfoxide and methanol.
Formula Weight 83.52
Percent Purity ≥98%
Chemical Name or Material Methoxylamine hydrochloride
compliance-icons compliance-icons
Product Identifier
  • Methoxylamine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Methoxylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.