missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Methoxyluteolin
GREENER_CHOICE

Product Code. 18735416
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18735416 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18735416 Supplier Tocris Bioscience™ Supplier No. 7320/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Inhibitor of pro-inflammatory mediator release from mast cells

Methoxyluteolin inhibits release of pro-inflammatory mediators from mast cells and decreases gene expression and release of TNF, VEGF and IL-8/CXCL8. The product also significantly inhibits IL-33 stimulated expression and release of CCL5 and CCL2. Methoxyluteolin pretreatment of LAD2 mast cells decreases gene expression of TNF, CXCL8 and VEGF in response to neurotensin or substance P via mTOR activation.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(3,4-Dimethoxyphenyl)-5,7-dimethoxy-4 H-1-benzopyran-4-one
CAS 855-97-0
Quantity 50 mg
Target Cytokine Compounds
Molecular Formula C19H18O6
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.