missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1,2,4-triazole-3-carboxylate, 98%

Product Code. 11456456
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456456 100 g 100g
11436456 5 g 5g
11446456 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456456 Supplier Thermo Scientific Alfa Aesar Supplier No. B24750.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4928-88-5 | C4H5N3O2 | 127.103 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4928-88-5
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.103
MDL Number MFCD00135989
InChI Key QMPFMODFBNEYJH-UHFFFAOYSA-N
PubChem CID 2735089
IUPAC Name methyl 1H-1,2,4-triazole-5-carboxylate
SMILES COC(=O)C1=NC=NN1

Specifications

Quantity 100 g
Synonym methyl 1h-1,2,4-triazole-3-carboxylate, methyl 1,2,4-triazole-3-carboxylate, 1h-1,2,4-triazole-3-carboxylic acid methyl ester, methyl 4h-1,2,4-triazole-3-carboxylate, 1,2,4-triazole-3-carboxylic acid methylester, 1h-1,2,4-triazole-3-carboxylic acid, methyl ester, 1h-1,2,4-triazole-5-carboxylic acid methyl ester, methyl1,2,4-triazole-3-carboxylate, acmc-20a2ir, 5-carbomethoxy-1,2,4-triazole
Formula Weight 127.1
Percent Purity 98%
Chemical Name or Material Methyl 1,2,4-triazole-3-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.