missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-cyclohexene-1-carboxylate, 97%

Product Code. 11467246
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467246 25 g 25g
11457246 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11467246 Supplier Thermo Scientific Alfa Aesar Supplier No. B25568.14
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18448-47-0 | C8H12O2 | 140.182 g/mol

Methyl 1-cyclohexene-1-carboxylate is used in the diastereoselective synthesis of cis-1,2-dialkenylcyclopropanols and methyl -7,7-dimethyl-9-oxo-1,3,4,4a,6,7,8,9,9b-decahydrodibenzo[b,d]furan-4a-carboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 1-cyclohexene-1-carboxylate is used in the diastereoselective synthesis of cis-1,2-dialkenylcyclopropanols and methyl -7,7-dimethyl-9-oxo-1,3,4,4a,6,7,8,9,9b-decahydrodibenzo[b,d]furan-4a-carboxylate.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and bases. 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18448-47-0
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.182
MDL Number MFCD00001544
InChI Key KXPWRCPEMHIZGU-UHFFFAOYSA-N
Synonym methyl 1-cyclohexene-1-carboxylate, methyl cyclohex-1-enecarboxylate, methyl 1-cyclohexenecarboxylate, methyl cyclohex-1-ene-1-carboxylate, 1-cyclohexene-1-carboxylic acid, methyl ester, cyclohexenecarboxylic acid, methyl ester, cyclohexene-1-carboxylic acid methyl ester, methyl1-cyclohexene-1-carboxylate, pubchem11031, 1-carbomethoxy cyclohexene
PubChem CID 87647
IUPAC Name methyl cyclohexene-1-carboxylate
SMILES COC(=O)C1=CCCCC1

Specifications

Density 1.03
Boiling Point 190°C to 192°C
Flash Point 73°C (165°F)
Refractive Index 1.477
Quantity 25 g
Solubility Information Immiscible with water.
Formula Weight 140.18
Percent Purity 97%
Chemical Name or Material Methyl 1-cyclohexene-1-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.