missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-hydroxy-1-cyclopropane carboxylate, 90%

Product Code. 10071831 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10071831 1 g Glass bottle 1g
10671642 5 g Glass bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10071831

Brand: Thermo Scientific Acros 302110010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33689-29-1 | C5H8O3 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33689-29-1
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD02093884
InChI Key KPJWVJURYXOHOO-UHFFFAOYSA-N
Synonym methyl 1-hydroxycyclopropanecarboxylate, methyl 1-hydroxy-1-cyclopropane carboxylate, methyl 1-hydroxy-1-cyclopropanecarboxylate, 1-hydroxycyclopropanecarboxylic acid methyl ester, 1-hydroxy-cyclopropanecarboxylic acid methyl ester, methyl-1-hydroxy-1-cyclopropane carboxylate, hydroxy-cyclopropanecarboxylic acid methyl ester, 1-hydroxy-cyclopropane-1-carboxylic acid methyl ester, cyclopropanecarboxylic acid, 1-hydroxy-, methyl ester
PubChem CID 2733178
IUPAC Name methyl 1-hydroxycyclopropane-1-carboxylate
SMILES COC(=O)C1(O)CC1

Specifications

Color Yellow
Boiling Point 70°C (13.0 mmHg)
Flash Point 17°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.4470 to 1.452
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in most organic solvents
Formula Weight 116
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Methyl 1-hydroxy-1-cyclopropane carboxylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.