missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-methyl-1H-imidazole-4-carboxylate, 97%, Thermo Scientific™

Product Code. 10350993
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10350993 1 g Amber Glass Bottle 1g
10681845 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10350993 Supplier Thermo Scientific Maybridge Supplier No. CC03622DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 17289-19-9
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD02179561
InChI Key KZPZTVKOJSKVBV-UHFFFAOYSA-N
Synonym methyl 1-methyl-1h-imidazole-4-carboxylate, 1-methyl-1h-imidazole-4-carboxylic acid methyl ester, 1h-imidazole-4-carboxylic acid, 1-methyl-, methyl ester, pubchem9002, acmc-209e5g, methyl1-methyl-1h-imidazole-4-carboxylate, ksc534i0d, methyl 1-methylimidazol-4-carboxylate, methyl 1-methyl-1h-imidazole-carboxylate, 4-methoxycarbonyl-1-methyl-1h-imidazole
PubChem CID 2773510
IUPAC Name methyl 1-methylimidazole-4-carboxylate
SMILES CN1C=C(N=C1)C(=O)OC

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-imidazole-4-carboxylic acid methyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.