missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-methylimidazole-5-carboxylate, 98%, Thermo Scientific™

Product Code. 11489890
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
Product Code. Quantity unitSize
11489890 0.25 g 0.25g
11499890 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11489890

Brand: Thermo Scientific Alfa Aesar 044688.01

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17289-20-2
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD01567300
InChI Key AKDPLDCXQNEMCL-UHFFFAOYSA-N
Synonym methyl 1-methyl-1h-imidazole-5-carboxylate, methyl 1-methylimidazole-5-carboxylate, methyl1-methyl-1h-imidazole-5-carboxylate, 3-methyl-3h-imidazole-4-carboxylic acid methyl ester, 1h-imidazole-5-carboxylic acid, 1-methyl-, methyl ester, 1-methyl-1h-imidazole-5-carboxylic acid methyl ester, pubchem9004, acmc-209e5h, 1-methylimidazole-5-carboxylic acid methyl ester, 1h-imidazole-5-carboxylicacid, 1-methyl-, methyl ester
PubChem CID 2736896
SMILES COC(=O)C1=CN=CN1C

Specifications

Melting Point 56°C to 58°C
Color Yellow
Quantity 0.25 g
IUPAC Name methyl 1-methyl-1H-imidazole-5-carboxylate
Formula Weight 140.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Methyl 1-methylimidazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.