missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-trifluoromethyl-1-cyclopropanecarboxylate, 96%, Thermo Scientific™

Product Code. 11349979
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349979 250 mg 250mg
11359979 1 g 1g
11369979 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11349979 Supplier Thermo Scientific Alfa Aesar Supplier No. H25757.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 208242-25-5
Molecular Formula C6H7F3O2
Molecular Weight (g/mol) 168.115
MDL Number MFCD06801321
InChI Key PAWUSHZXEZPDBE-UHFFFAOYSA-N
Synonym methyl 1-trifluoromethyl cyclopropane-1-carboxylate, 1-trifluoromethyl cyclopropane-1-carboxylic acid methyl ester, methyl 1-trifluoromethyl cyclopropanecarboxylate, methyl 1-trifluoromethyl-1-cyclopropanecarboxylate, 1-trifluoromethylcyclopropane-1-carboxylic acid methyl ester, acmc-1cj3i, 1-trifluoromethyl cyclopropane-1-carboxylic acidmethyl ester, 1-trifluoromethyl-cyclopropane-1-carboxylic acid methyl ester
PubChem CID 24208784
IUPAC Name methyl 1-(trifluoromethyl)cyclopropane-1-carboxylate
SMILES COC(=O)C1(CC1)C(F)(F)F

Specifications

Boiling Point 115°C
Flash Point 42°C (108°F)
Quantity 250 mg
UN Number UN3272
Formula Weight 168.12
Percent Purity 96%
Chemical Name or Material Methyl 1-trifluoromethyl-1-cyclopropanecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.